Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents
Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents
Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents
Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents
Glycerol Gel-Loading Dye, 5X (Contains 30% Glycerol/Molecular Biol.), Fisher BioReagents
Have Questions?
Catalog NumberQuantityPackaging
FBRBP645-55 mLPoly Bottle
Catalog number FBRBP645-5
Price (MYR)
-
Quantity:
5 mL
Packaging:
Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS56-81-5, 7732-18-5
Specifications
Chemical Name or MaterialGlycerol Gel-Loading Dye, 5X
Name NoteContains 30% Glycerol
CAS56-81-5
ChemAlert Storage SymbolGray
Absorbance0.25 to 0.40 AU, 0.25 to 0.45 AU at 252nm, 0.30 to 0.45 AU
View more
This dye is added to DNA and RNA samples prior to electrophoresis on agarose gels.