Check Order Status
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Calcium Chloride Dihydrate (White Crystals to Powder), Fisher BioReagents™
Have Questions?
Change view
Quantity:
500 g
100 g
250 g
Packaging:
Poly Bottle
Catalog number
FBRBP510-500
Price (MYR)
424.00
EA
Add to cart
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
424.00
EA
Add to cart
Specifications
Chemical Name or Material
Calcium Chloride Dihydrate
Melting Point
175°C
CAS
10035-04-8
ChemAlert Storage Symbol
Gray
Color
Beige-White
View more
Calcium Chloride Dihydrate (White Crystals to Powder)
Calcium Chloride is suitable for use in the preparation of buffer solutions.