Check Order Status
Sorbitol, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Sorbitol, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP439-500
500 g
Catalog number
FBRBP439-500
Price (MYR)
524.00
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
524.00
Each
Add to cart
Specifications
Chemical Name or Material
Sorbitol
CAS
50-70-4
ChemAlert Storage Symbol
Gray
Melting Point
75°C
Color
White
View more
Sorbitol is used in the preparation of isoelectric focusing gels.