Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Fisher Chemical
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
Have Questions?
Catalog NumberQuantityPackaging
FBRBP381-11 kgPoly Bottle
Catalog number FBRBP381-1
Price (MYR)
608.00
Each
Add to cart
Quantity:
1 kg
Packaging:
Poly Bottle
Request bulk or custom format
Price (MYR)
608.00
Each
Add to cart
Specifications
Chemical Name or MaterialGlycine
Melting Point182°C
Absorbance0.1 AU max. at 280nm
Ammonium0.02% max.
CAS56-40-6
View more
Glycine (White Crystals or Crystalline Powder)
Glycine is used in the preparation of Tris–Glycine electrophoresis buffers and culture media.