Check Order Status
L-Cysteine Hydrochloride Monohydrate (White Crystals), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
L-Cysteine Hydrochloride Monohydrate (White Crystals), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP376-100
100 g
Catalog number
FBRBP376-100
Price (MYR)
661.00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
661.00
Each
Add to cart
Specifications
Chemical Name or Material
L-Cysteine Hydrochloride Monohydrate
Melting Point
175°C
Ammonium
0.02% max.
CAS
4-6-7048
ChemAlert Storage Symbol
Gray
View more
This amino acid is suitable for use in tissue culture systems requiring additives.