L-Arginine Hydrochloride (White Crystals or Crystalline Powder), Fisher BioReagents
L-Arginine Hydrochloride (White Crystals or Crystalline Powder), Fisher BioReagents
L-Arginine Hydrochloride (White Crystals or Crystalline Powder), Fisher BioReagents
L-Arginine Hydrochloride (White Crystals or Crystalline Powder), Fisher BioReagents
Fisher Chemical
L-Arginine Hydrochloride (White Crystals or Crystalline Powder), Fisher BioReagents
Have Questions?
Catalog NumberQuantity
FBRBP372-100100 g
Catalog number FBRBP372-100
Price (MYR)
481.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
481.00
EA
Add to cart
Specifications
Chemical Name or MaterialL-Arginine Hydrochloride
Melting Point226°C
Ammonium0.02% max.
CAS1119-34-2
ChemAlert Storage SymbolGray
View more
The hydrochloride salt of L-Arginine is suitable for use in tissue culture systems requiring additives and has many of the same applications as L-Arginine.