Check Order Status
Potassium Phosphate Monobasic (White Crystals), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Potassium Phosphate Monobasic (White Crystals), Fisher BioReagents
Have Questions?
Change view
Quantity:
500 g
Packaging:
Glass Bottle
Catalog number
FBRBP362-500
Price (MYR)
510.00
Each
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
510.00
Each
Add to cart
Specifications
Chemical Name or Material
Potassium Phosphate Monobasic
Melting Point
253°C
CAS
7778-77-0
ChemAlert Storage Symbol
Gray
Color
White
View more
This potassium salt is used in buffers for molecular biology and cell culture applications.