Check Order Status
Sodium Hydroxide (White Pellets), Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
Sodium Hydroxide (White Pellets), Fisher BioReagents™
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change view
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Catalog number
FBRBP359-500
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Poly Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Sodium Hydroxide
pH
14
Color
White
Boiling Point
1390°C
CAS
1310-73-2
View more
Sodium Hydroxide (White Pellets)
Sodium Hydroxide is used to adjust the pH of buffers for molecular biology and cell culture applications.