Check Order Status
CAPS (Fine White Crystals), Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
CAPS (Fine White Crystals), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP321-100
100 g
Glass Bottle
Catalog number
FBRBP321-100
Price (MYR)
664.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
664.00
EA
Add to cart
Specifications
Chemical Name or Material
CAPS
CAS
1135-40-6
ChemAlert Storage Symbol
Gray
Melting Point
324°C
Color
White
View more
This biological buffer has a usable pH range of 9.7 to 11.1.