Check Order Status
Nystatin, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
Nystatin, Fisher BioReagents
Alters membrane permeability and allows potassium ion leakage
Have Questions?
Catalog Number
Quantity
FBRBP2949-5
5 g
Catalog number
FBRBP2949-5
Price (MYR)
620.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
620.00
EA
Add to cart
Specifications
Chemical Name or Material
Nystatin
Type
Nystatin
Physical Form
Solid
CAS
1400-61-9
Health Hazard 2
CAUTION!
View more