Water, Molecular Biology Grade, Fisher BioReagents
Water, Molecular Biology Grade, Fisher BioReagents
Water, Molecular Biology Grade, Fisher BioReagents
Water, Molecular Biology Grade, Fisher BioReagents
Water, Molecular Biology Grade, Fisher BioReagents
Fisher Chemical
Water, Molecular Biology Grade, Fisher BioReagents
For many fundamental molecular biology procedures
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
100 mL
1 L
Packaging:
View all
Poly CUBE
Catalog number FBRBP2819-4
Price (MYR)
576.00
EA
Add to cart
Quantity:
4 L
Packaging:
Poly CUBE
Request bulk or custom format
Price (MYR)
576.00
EA
Add to cart
Specifications
Chemical Name or MaterialWater
Name Note0.03μm filtered to ensure high purity
pH7
CAS7732-18-5
ChemAlert Storage SymbolGray
View more
Ultrapure, deionized, molecular biology grade water is ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses.