RiboLadder™ RNA Standards
RiboLadder™ RNA Standards
Actual product may vary
RiboLadder™ RNA Standards
For assessment of single-stranded RNA molecules on both native or denaturing agarose and polyacrylamide gels
Have Questions?
Catalog NumberNo. of BandsRange
FBRBP2810-508 0.1 to 1 kb
Catalog number FBRBP2810-50
Price (MYR)
-
No. of Bands:
8
Range:
0.1 to 1 kb
  • Genetically engineered for precise bands
  • Well-spaced bands for easy identification and sizing of unknown RNA fragments and higher intensity reference band for fast size comparison
  • Packaged in two vials (25 loadings each) and lyophilized to decrease thawing-related degradation, to prolong shelf life and to ensure consistent performance
  • Purity level suitable for capillary electrophoresis and bioanalyzer applications
  • Two size ranges suitable for most RNA applications
  • Ideal for ethidium bromide and fluorescent staining
Specifications
Description100 bp RNA standard
ColorBlue
Product TypeRNA Molecular Weight Marker/Ladder
FormLyophilized
For Use With (Application)Assessment of single stranded RNA molecules on both native or denaturing Agarose and Polyacrylamide gels
Kit ContentsTwo vials of RNA standard (25 Dye loads each), 1X loading Buffer that contains Bromophenol Blue which serves as a tracking dye and 2X sample loading Buffer
No. of Bands8
Quantity50 Loadings
Range 0.1 to 1 kb
Storage Requirements−70°C
Unit SizeEA