▸
Gel Electrophoresis Equipment and Supplies
▸
Gel Electrophoresis Reagents and Buffers
▸
Nucleic Acid Molecular Weight Markers
▸
ex
ACTG
ene
™ DNA Ladders
Actual product may vary
ex
ACTG
ene
™ DNA Ladders
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
Have Questions?
Catalog Number
For Use With (Application)
No. of Bands
FBRBP2579-100
General purpose, wide size range
19
Catalog number
FBRBP2579-100
Price (MYR)
-
For Use With (Application):
General purpose, wide size range
No. of Bands:
19
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
Ideal for size assessment and quantitative determination of DNA mass
Contain loading dye to reduce pipetting steps and save time
Have two or three high intensity reference bands for better sizing
Provided in sufficient quantity to load 100 lanes
Specifications
Color
Colorless
Form
Liquid
Formulation
Premixed with loading dye
For Use With (Application)
General purpose, wide size range
No. of Bands
19
Quantity
100 Lanes
Storage Requirements
Stable at room temperature for two years.
Sufficient For
100 Lanes
Product Type
DNA Molecular Weight Marker