Check Order Status
D-(-)-Arabinose (White Powder), Fisher BioReagents
Element Search
Structure Search
D-(-)-Arabinose (White Powder), Fisher BioReagents
Have Questions?
Catalog Number
Quantity
FBRBP2504-100
100 g
Catalog number
FBRBP2504-100
Price (MYR)
-
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material
D-(-)-Arabinose
CAS
28697-53-2
ChemAlert Storage Symbol
Gray
Melting Point
152°C
Color
White
View more
D-(-)-Arabinose is a component used for culture media.