Check Order Status
TBS, Tris Buffered Saline, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
TBS, Tris Buffered Saline, 1X Solution, pH 7.4, Molecular Biology, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP2472-1
1 L
Nalgene Poly Bottle
Catalog number
FBRBP2472-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7647-14-5, 7447-40-7, 7647-01-0, 7732-18-5, 77-86-1
Specifications
Chemical Name or Material
Tris Buffered Saline
Name Note
1X Solution, pH 7.4
CAS
7647-14-5
ChemAlert Storage Symbol
Gray
Boiling Point
100°C
View more
Tris Buffered Saline (TBS) is used in biochemical and molecular biology applications.