Check Order Status
D-Biotin, Fisher BioReagents
Element Search
Structure Search
Fisher Chemical
D-Biotin, Fisher BioReagents
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP232-1
1 g
Amber Glass
Catalog number
FBRBP232-1
Price (MYR)
737.00
EA
Add to cart
Quantity:
1 g
Packaging:
Amber Glass
Request bulk or custom format
Price (MYR)
737.00
EA
Add to cart
Specifications
Alpha Vector
Biotin
Chemical Name or Material
D-Biotin
CAS
58-85-5
ChemAlert Storage Symbol
Gray
Health Hazard 2
CAUTION!
View more
D-Biotin, which forms a stable complex with Avidin, is useful in the study of Avidin-Biotin interactions and in affinity chromatography.