▸
Solvents
▸
Other Solvents
▸
Ethylene Glycol
▸
Ethylene Glycol, Fisher BioReagents™
Element Search
Structure Search
Ethylene Glycol, Fisher BioReagents™
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP230-1
1 L
Glass Bottle
Catalog number
FBRBP230-1
Price (MYR)
-
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Material
Ethylene Glycol
pH
5.5 to 7.5
Melting Point
-13°C
CAS
107-21-1
ChemAlert Storage Symbol
Gray
View more
Ethylene Glycol
Due to its property as an antifreeze, Ethylene Glycol may be used for the storage of enzymes at low temperatures.