Microbiology Media: LB Broth, Miller
Microbiology Media: LB Broth, Miller
Actual product may vary
Microbiology Media: LB Broth, Miller
Used for growing molecular biology strains of E. coli-containing recombinant plasmids
Have Questions?
Catalog NumberQuantity
FBRBP1426-500500 g
Catalog number FBRBP1426-500
Price (MYR)
-
Quantity:
500 g
Dehydrated.
Specifications
FormPowder
GradeMolecular genetics
Media TypeLB Broth
Product TypeMicrobiology Media
FormulationTryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
For Use With (Application)For preparing bacterial nutrient media
Packaging TypePoly Bottle
Quantity500 g
Unit SizeBOT
Contents & Storage
4°C