Check Order Status
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology, Fisher BioReagents™
Element Search
Structure Search
Fisher Chemical
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology, Fisher BioReagents™
Have Questions?
Catalog Number
Quantity
Packaging
FBRBP1338-1
1 L
Nalgene Poly Bottle
Catalog number
FBRBP1338-1
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
77-86-1, 60-00-4, 7732-18-5
Specifications
Chemical Name or Material
Tris-EDTA
Name Note
100X Solution
CAS
77-86-1
ChemAlert Storage Symbol
Gray
Color
Undesignated
View more
TE Buffer, Tris-EDTA, 100X Solution, pH 8.0, Molecular Biology
Tris–EDTA (TE) is routinely used for suspending nucleic acid samples.