Isoamyl Alcohol (Molecular Biology), Fisher BioReagents
Actual product may vary
Isoamyl Alcohol (Molecular Biology), Fisher BioReagents
Isoamyl Alcohol (Molecular Biology), Fisher BioReagents
Isoamyl Alcohol (Molecular Biology), Fisher BioReagents
CAS: 123-51-3 | C5H12O | 88.15 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FBRBP1150-500500 mLGlass Bottle
Catalog number FBRBP1150-500
Price (MYR)
-
Quantity:
500 mL
Packaging:
Glass Bottle
500mL; Glass Bottle
A mixture of Chloroform and Isoamyl Alcohol (24:1 v/v) is used to remove proteins during nucleic acid extraction procedures. It may also be used to extract Ethidium Bromide from nucleic acids.
Specifications
Chemical Name or MaterialIsoamyl Alcohol
Cleanup SymbolSolvent
Acidity0.01% max.
CAS123-51-3
ChemAlert Storage SymbolRed
Health Hazard 2WARNING!
Health Hazard 3Emergency Overview
Flammable liquid and vapor. Irritating to eyes and respiratory system. Aspiration hazard if swallowed - can enter lungs and cause damage. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.
NFPA
Health:1
Flammability:2
Instability:0
Melting Point-117°C
Recommended StorageRT
pH6.5
Boiling Point130°C
Assay Percent Range≥99 %
PackagingGlass Bottle
Molecular FormulaC5H12O
Linear Formula(CH3)2CHCH2CH2OH
Quantity500 mL
Residue after Evaporation0.003% max.
Merck Index15, 5241
Synonym3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
InChI KeyPHTQWCKDNZKARW-UHFFFAOYSA-N
SMILESCC(C)CCO
IUPAC Name3-methylbutan-1-ol
Molecular Weight (g/mol)88.15
PubChem CID31260
ChEBICHEBI:15837
Formula Weight88.15
Vapor Pressure4hPa at 20°C
Percent Purity≥99%
GradeMolecular Biology
Physical FormLiquid
Unit SizeEach