Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Fisher Chemical
Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Packaging:
Glass Bottle
Catalog number FBRBP1105-4
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethanol
Cleanup SymbolSolvent
Melting Point-98°C
Name NotePeroxide-free
Absorbance0.025 max. at 254nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
View more
Methanol (Peroxide-Free/Sequencing)
Methanol is used in staining and destaining protein electrophoresis gels, in HPLC, and in other biological applications.