Elmasonic X-tra ST Heated Ultrasonic Cleaners
Elmasonic X-tra ST Heated Ultrasonic Cleaners
Actual product may vary
Elmasonic X-tra ST Heated Ultrasonic Cleaners
Elmasonic X-tra ST Heated Ultrasonic Cleaners
Elmasonic X-tra ST Heated Ultrasonic Cleaners
The Elmasonic X-tra ST Heated Ultrasonic Water Baths are designed for use in a rugged production environment, workshops and servicing. Designed for continuous operation at the highest load levels, in single-shift operation with a three-year warranty.
Have Questions?
Catalog NumberCapacity (Metric)Dimensions (D x W x H) InteriorVoltage
FBE1076440126 L (Working volume: 97 L)600 x 600 x 350 mm3 x 400 V
Catalog number FBE1076440
Price (MYR)
-
Capacity (Metric):
126 L (Working volume: 97 L)
Dimensions (D x W x H) Interior:
600 x 600 x 350 mm
Voltage:
3 x 400 V
  • Ultrasonic trough made of stainless steel
  • Sloping trough base to drain the cleaning liquid better
  • Sandwich high-performance oscillation systems
  • Ultrasound frequency can be switched over between 25 kHz for coarse and pre-cleaning and 45 kHz for fine cleaning
  • Sweep function for continuous shifting of the maximum sound pressures and bringing about more homogenous sound distribution in the cleaning bath
  • Switchable pulse function to intensify the ultrasonic cleaning effect; in addition, optimization of readiness for operation (reaching the cavitation threshold) and after replacing the basket (reduction in cleaning time)
  • Dynamic function: the sweep and pulse functions run alternately on an automatic basis; the ultrasound output is increased by up to 20%
  • At the same time, the uniform distribution of ultrasound in the ultrasonic bath enhances the cleaning effect
  • Heating with temperature regulation from 30° to 80°C
  • Temperature-controlled ultrasound function; the ultrasound starts automatically when the preselected temperature is reached
  • Automatic safety shut-down after 12 hours of operation to avoid accidental continuous operation
  • Automatic safety shut-down at bath temperature of 90°C to protect from excessive temperature
  • Display of the set values and the actual values on the LED display
  • Housing made of stainless steel
  • Rollers with a parking brake to move the device when it is empty
Specifications
MaterialStainless Steel
DisplayLED
Voltage3 x 400 V
Capacity (Metric)126 L (Working volume: 97 L)
Dimensions (D x W x H)850 x 885 x 936 mm
Dimensions (D x W x H) Interior600 x 600 x 350 mm
For Use With (Application)Ultrasonic cleaning
Model1400H
Wattage6800 w
Weight (Metric)87 kg
Operating ModesSweep, Pulse, Dynamic
Operating Frequency25/45 kHz
Unit SizeEach