Check Order Status
Stainless Steel Baskets for Elmasonic Xtra ST Ultrasonic Cleaner
Actual product may vary
Stainless Steel Baskets for Elmasonic Xtra ST Ultrasonic Cleaner
For use with Elmasonic Xtra ST Ultrasonic Cleaner.
Have Questions?
Catalog Number
For Use With (Equipment)
Dimensions (L x W x H)
FBE1027350
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H
49 x 52.5 x 20.8 cm
Catalog number
FBE1027350
Price (MYR)
-
For Use With (Equipment):
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H
Dimensions (L x W x H):
49 x 52.5 x 20.8 cm
Holds parts when immersing and removing from solution
Open weave in sides and bottoms
Protect cleaner and parts from damage caused by direct contact
Specifications
Type
Basket
For Use With (Equipment)
Ultrasonic Cleaner, Elmasonic Xtra ST 1400 H
For Use With (Application)
Hold Parts When Immersing and Removing Them From Solution in Ultrasonic Cleaners
Material
Stainless Steel
Dimensions (L x W x H)
49 x 52.5 x 20.8 cm