P-Series Ultrasonic Cleaner
P-Series Ultrasonic Cleaner
Actual product may vary
P-Series Ultrasonic Cleaner
Fisherbrand™ P-Series Ultrasonic Water Baths have convenient digital display of set and operating parameters, a cavitation resistant stainless steel tank, ergonomic handles for easy positioning and a durable lid that helps reduce sound and solution evaporation.
Have Questions?
Catalog NumberCapacity (Metric)ModelDimensions (D x W x H)
FBE101380418 LP 180 H340 x 390 x 321 mm
Catalog number FBE1013804
Price (MYR)
-
Capacity (Metric):
18 L
Model:
P 180 H
Dimensions (D x W x H):
340 x 390 x 321 mm
For all applications in analytical, industrial and medical laboratories, including intensive cleaning applications.
  • Significantly more powerful than conventional ultrasonic units
  • Practical and easy to operate with clear display showing set and actual parameters
  • Multi-frequency
  • 37kHz for standard applications
  • 80kHz for prolonged cleaning time, ideal for the treatment of very small and sensitive items
  • Units feature normal, pulse, sweep, degas and power control functions
  • Side-mounted drain valve and reasr drain duct to remove spent solution
  • Powerful pulse mode removes stubborn contaminants
  • Stainless steel basket, plastic cover available separately
Specifications
MaterialV2A Stainless Steel
Voltage230 VAC
Capacity (Metric)18 L
Dimensions (D x W x H)340 x 390 x 321 mm
Electrical Requirements230VAC, 1400w
For Use With (Application)Professional applications in analytical and medical laboratories and in industrial production processes
ModelP 180 H
Temperature (Metric) Range5°C to 40°C
Weight (Metric)8.5 kg
Unit SizeEach