PEEK Fingertight Fittings
PEEK Fingertight Fittings
Actual product may vary
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
PEEK Fingertight Fittings
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDescription
F-130XOne-Piece PEEK Long Fingertight Fitting1/16 in., 0.37 in. Head
F-120X
also known as IDEX = F-120X
One-Piece PEEK Fingertight Fitting1/16 in., 0.37 in. Head
F-300X
also known as 00101-18195
PEEK Wing Nut w/PEEK Ferrule1/16 in.
HYPU-467BLKShort Head Port Plug, Black1/16 in., 10-32 Coned, Delrin, Black
U-467BLKShort Head Port Plug, Black1/16 in., 10-32 Coned, Delrin, Black
Catalog number F-130X
Price (MYR)
-
Type:
One-Piece PEEK Long Fingertight Fitting
Description:
1/16 in., 0.37 in. Head
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
Description
  • Resist cracking, breaking, thread stripping and leaking in both low and high pressure applications
  • Biocompatible for a broad range of applications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description1/16 in., 0.37 in. Head
For Use With (Equipment)LC Tubing, LC Tubing
MaterialPEEK, PEEK
TypeOne-Piece PEEK Long Fingertight Fitting
Quantity10 Pack
Length (English)1.03 in.
Pressure Rating6000 psi