Dexfenfluramine hydrochloride
Dexfenfluramine hydrochloride
Actual product may vary
Dexfenfluramine hydrochloride
5-HT reuptake inhibitor; also stimulates 5-HT release
Have Questions?
Catalog NumberQuantity
EYE26951010 mg
Catalog number EYE269510
Price (MYR)
600.00
Special offer
754.00
Save 154.00 (20%)
EA
Add to cart
Quantity:
10 mg
Price (MYR)
600.00
Special offer
754.00
Save 154.00 (20%)
EA
Add to cart
Indirectly agonizes serotonin receptors via inhibition of 5-HT re-uptake and stimulation of 5-HT release. Anorectic agent; decreases growth hormone, insulin, leptin, fat mass, lean mass and increases ghrelin in diet-switched diet-induced obese mice.
Specifications
Chemical Name or Material(S)-N-Ethyl-α-methyl-3-(trifluoromethyl)benzeneethanamine hydrochloride
CAS3239-45-0
Recommended StorageDesiccate at RT
Quantity10 mg
Target5-HT Transporter Inhibitors
Molecular FormulaC12H16F3N.HCl
Purity0.95
Unit SizeEA