JP 1302 dihydrochloride
JP 1302 dihydrochloride
JP 1302 dihydrochloride
JP 1302 dihydrochloride
JP 1302 dihydrochloride
Potent and selective α2C antagonist
Have Questions?
Catalog NumberQuantity
EYE26665050 mg
Catalog number EYE266650
Price (MYR)
18,835.00
EA
Add to cart
Quantity:
50 mg
Request bulk or custom format
Price (MYR)
18,835.00
EA
Add to cart
Specifications
Chemical Name or MaterialJP 1302 dihydrochloride
CAS1259314-65-2
Recommended StorageStore at -20°C
Quantity50 mg
Molecular FormulaC24H26Cl2N4
View more
α2C-adrenoceptor antagonist that displays ∽ 50-fold selectivity over other α2-adrenoceptor subtypes (Ki values are 28, 1470, 1700 and 3150 nM for human α2C, α2B, α2D and α2A subtypes respectively). Potently antagonizes adrenalin-stimulated 35GTPγS binding in vitro (KB = 16 nM) and produces antidepressant and antipsychotic-like effects in vivo.