RODAC™ SNAP LID Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 (LP80)
RODAC™ SNAP LID Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 (LP80)
Actual product may vary
RODAC™ SNAP LID Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 (LP80)
RODAC™ SNAP LID Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 (LP80)
RODAC™ SNAP LID Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 (LP80)
Have Questions?
Catalog NumberQuantityPackaging TypeShelf Life
EYE21527010/Pk.VHP Resistant Triple Wrapping Isolator Pack112 Days
Catalog number EYE215270
Price (MYR)
242.00
EA
Add to cart
Quantity:
10/Pk.
Packaging Type:
VHP Resistant Triple Wrapping Isolator Pack
Shelf Life:
112 Days
Price (MYR)
242.00
EA
Add to cart
The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.
The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:
  • Withstands an accidental drop from the height of a bench-top workspace
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage
Specifications
Certifications/ComplianceISO/DIS, 11014
ColorLight Beige
FormLiquid
Manufacturing OriginUnited States
Media TypeSabouraud Dextrose Agar with Lecithin and Polysorbate 80
SterilitySterile
IncludesRodac™ snap lid
Packaging TypeVHP Resistant Triple Wrapping Isolator Pack
Quantity10/Pk.
Shelf Life112 Days
Unit SizeEA