(S)-(+)-Dimethindene maleate
(S)-(+)-Dimethindene maleate
(S)-(+)-Dimethindene maleate
(S)-(+)-Dimethindene maleate
(S)-(+)-Dimethindene maleate
M2-selective antagonist
Have Questions?
Catalog NumberQuantity
EYE14251010 mg
Catalog number EYE142510
Price (MYR)
758.00
EA
Add to cart
Quantity:
10 mg
Request bulk or custom format
Price (MYR)
758.00
EA
Add to cart
Specifications
Chemical Name or Material(S)-(+)-Dimethindene maleate
CAS136152-65-3
Recommended StorageDesiccate at Room Temperature
Quantity10 mg
Molecular FormulaC24H28N2O4
View more
(S)-(+)-Dimethindene maleate is an enantiomer that is a subtype-selective M2 muscarinic receptor antagonist (pKi values are 7.08, 7.78, 6.70 and 7.00 for M1, M2, M3 and M4 receptors respectively). Also H1 histamine receptor antagonist (pKi = 7.48). Allows formation of extended pluripotent stem (EPS) cells in combination with CHIR 99021 (Cat.No. 4423), minocycline hydrochloride (Cat.No. 3268) and human leukemia inhibitory factor.
For more information about how Dimethindene maleate may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .