Search Thermo Fisher Scientific
Reduce accumulation of glass chips within instrumentation using Thermo Scientific™ Superfrost™ Plain and Frosted Clipped Corner Microscope Slides.
Catalog Number | Description |
---|---|
ERW29514 | Sandblasted |
Reduce accumulation of glass chips within instrumentation using Thermo Scientific™ Superfrost™ Plain and Frosted Clipped Corner Microscope Slides. Ideal for histology, hematology and microbiology applications, the 45° clipped corners enable ease of handling.