Electrothermal™ Powder Coated Aluminum Slide Drying Bench
Electrothermal™ Powder Coated Aluminum Slide Drying Bench
Actual product may vary
Electrothermal™ Powder Coated Aluminum Slide Drying Bench
Aids in the preparation of microscope slides at the specimen mounting stage. Bibby Scientific™ Electrothermal™ accepts up to 50 slides and has the facility for drying slides in different orientations.
Have Questions?
Catalog NumberProduct Type
ELEMH6616Slide Drying Bench
Catalog number ELEMH6616
Price (MYR)
9,161.00
EA
Add to cart
Product Type:
Slide Drying Bench
Price (MYR)
9,161.00
EA
Add to cart
Aids in the preparation of microscope slides at the specimen mounting stage. Bibby Scientific™ Electrothermal™ accepts up to 50 slides and has the facility for drying slides in different orientations.
The bestselling Slide Drying Bench aids in the preparation of microscope slides at the specimen mounting stage. It complements both the Paraffin Wax Dispenser and the Paraffin Wax Mounting Bath to provide a complete set of paraffin wax sampling equipment for the modern laboratory. Accepting up to 50 slides, it has the facility for drying slides in different orientations- across the drying support bars, angled from the bars, or flat without the bars.
  • Up to 50 slides capacity
  • The slides can be arranged on the bench in different orientations
  • Built-in energy regulator controlling temperature up to 100°C
Specifications
Product TypeSlide Drying Bench
Slide Capacity50
MaterialPowder-coated Aluminum
For Use With (Application)Drying of multiple microscope slides at the specimen mounting stage, and complements the Electrothermal Paraffin Wax Dispenser and the Electrothermal Paraffin Wax Mounting Bath in the provision of an ideal set of sample preparation equipment for the modern histology laboratory
IncludesUK and EU plugs
Quantity1 Ea.
Temperature Range (Metric)10°C to 100°C
Depth (Metric)180 mm
Height (Metric)95 mm
Width (Metric)390 mm
Voltage230 V
Wattage150 w
Unit SizeEA