Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
Actual product may vary
Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
Savant™ SpeedVac™ DNA 130 Integrated Vacuum Concentrator System
Save time and money with these Concentrator Kits. Everything you need for quick drying and concentrating of small sample volumes
Have Questions?
Catalog NumberDescriptionElectrical Requirements
DNA130-230SpeedVac DNA130 Vacuum Concentrator System230 V 50 Hz
Catalog number DNA130-230
Price (MYR)
73,038.00
EA
Add to cart
Description:
SpeedVac DNA130 Vacuum Concentrator System
Electrical Requirements:
230 V 50 Hz
Price (MYR)
73,038.00
EA
Add to cart
Designed for fast, efficient concentration of DNA and RNA applications, the Thermo Scientific™ Savant SpeedVac™ DNA130 is a compact, integrated vacuum concentrator. SpeedVac™ DNA130 vacuum concentrator will support sample preparations with ease and efficiency.
  • Compact, easy to use design: smallest integrated benchtop unit saves lab bench space.
  • Corrosion-resistant, PTFE coated chamber offers years of service
  • Built-in, oil free vacuum pump offers maintenance-free operation
  • Includes a rotor that holds 36 x 1.5-2.0 ml tubes (RD36)
Operating Features:
  • Consistent performance: designed to ensure consistent drying times and reproducible results
  • User-friendly interface
  • Three user defined programs that can be modified and saved
  • Temperature set point: ambient, 35°C to 65°C in 5°C increments
  • Heat Time setting: from 1 m to 9h 59 m or continuous
  • Run Time setting: from 1m to 9h 59m
  • Audible alarm to alert user when run is complete
Sample Protection
  • Dry at ambient temperatures or using low heat to avoid sample denaturing
  • Oil-free diaphragm pump provides gentle transition from atmosphere to ultimate vacuum setting with no solvent bumping
Safety Features
  • PTFE-coated chamber resists chemicals and cleans easily
  • Latched lid and lid lock indicator ensures user safety during operation
  • Lid stay mechanism to secure lid while it‘s open, allowing user to have both hands free for operation
  • Unit and pump are virtually maintenance-free
  • External post trap to neutralize ammonium hydroxide vapors
  • Optional chemical traps and optional cartridges to trap radioactive chemicals to minimize concerns in handling hazaRDO™us fumes and chemicals.
Solvents and Solvent Combinations:
  • Water
  • PCR buffers (aqueous)
  • Ethanol and water
  • Ammonium Hydroxide (will require an ammonia post-trap ANT-100 and neutralizing solution accessory ANS121)
Specifications
CertificationsUL Listed to US and Canadian requirements. CE marked
HoldsAccommodates 36 x 1.5 mL Tubes, 72 x 0.5 mL Tubes and 2 Microwell Plates
IncludesRD36 rotor (36x 1.5-2.0 ml tubes)
Max. Vacuum<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)105 lb.
Shipping Weight (Metric)48 kg
Voltage230 V
Temperature Range (Metric)Ambient to 65°C
Chamber MaterialPTFE-coated Aluminum
DescriptionSpeedVac DNA130 Vacuum Concentrator System
Electrical Requirements230 V 50 Hz
Amperage6 A
Plug TypeSchuko CEE 7-7
Weight (English)86 lb.
Weight (Metric)39 kg
Dimensions (L x W x H)11.5 x 25 x 12 in. (29 x 64 x 31 cm)
Unit SizeEA