Decon 90
Decon 90
Actual product may vary
Decon 90
Biodegradable, phosphate and enzyme free, water rinsable surface active cleaning agent and radioactive decontaminant
Have Questions?
Catalog NumberVolume (Metric)
DECD002521
also known as DECD/0025/21
5000 mL
Catalog number DECD002521
also known as DECD/0025/21
Price (MYR)
260.00
BOT
Add to cart
Volume (Metric):
5000 mL
Price (MYR)
260.00
BOT
Add to cart
Biodegradable, phosphate and enzyme free, water rinsable surface active cleaning agent and radioactive decontaminant
Biodegradable, phosphate and enzyme free, water rinsable surface active cleaning agent and radioactive decontaminant. A 2% to 5% solution is adequate for the majority of cleaning applications. Increasing solution strength, heating the solution or use in an ultrasonic cleaning bath will greatly reduce the cleaning time. Not suitable for use on non-ferrous metals, notably aluminum and zinc, or on polycarbonate. Concentrate pH 13.0, 10% solution pH 11.9, 2% solution pH 10.7. Ideal for laboratory, medical and specialised industrial cleaning applications.
Specifications
FormLiquid
pH>13
AppearanceWhite
CompositionPhosphate, Enzyme-Free, Water, 2% to 5% Solution
Melting Point-3.8°C
Viscosity6.27cps at 20°C
Volume (Metric)5000 mL
Unit SizeBOT