Barnstead™ Nanopure™ Accessories and Consumables
Barnstead™ Nanopure™ Accessories and Consumables
Actual product may vary
Barnstead™ Nanopure™ Accessories and Consumables
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
D50253Cartridge, Ultrapure Deionization
D50280R/O and Distilled Feed (Ultralow Organics, Type I Water)
D50251Cartridge, Pretreatment (Tap and DI Water)
D50256Pretreatment Cartridge Kit (Tap and DI Water)
D50281
also known as BARNSTEAD = D50281
Deionized Water Feed (Ultralow Organics, Type I Water)
D50282R/O and Distilled Feed (Low Organics, Type I Water)
D50283Deionized Water Feed (Low Organics, Type I Water)
Catalog number D50253
Price (MYR)
1,517.00
EA
Add to cart
Type:
Cartridge, Ultrapure Deionization
Price (MYR)
1,517.00
EA
Add to cart
Optimize your water purification system with Thermo Scientific™ Barnstead™ Nanopure™ Accessories and Consumables. Choose the cartridge pack according to your feed water and system. Cartridge packs ship with 0.2 micron final filter. Other products include UV lamp and low water level pump protector.
  • Simple, one-connection installation and removal
Includes: One 0.2μm absolute final filter
Specifications
For Use With (Equipment)Nanopure Infinity System
TypeCartridge, Ultrapure Deionization
Unit SizeEA