Check Order Status
Barnstead™ Deionization System
Actual product may vary
Barnstead™ Deionization System
For the Nanopure, E-pure Water Systems
Have Questions?
Catalog Number
Type
D5021
Deionization Cartridge
Catalog number
D5021
Price (MYR)
1,517.00
EA
Add to cart
Type:
Deionization Cartridge
Price (MYR)
1,517.00
EA
Add to cart
Deionization System
For the Nanopure, E-pure Water Systems
Specifications
Pore Size
60 μm
Resin Type
Mixed Bed
For Use With (Application)
Deionization System
For Use With (Equipment)
Quad, E-Pure, Deionization Cartridge
Type
Deionization Cartridge