Barnstead™ Cartridge Kits
Barnstead™ Cartridge Kits
Actual product may vary
Barnstead™ Cartridge Kits
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Have Questions?
Catalog NumberFor Use With (Equipment)Feedwater SourceIncludesModel
D3804Nanopure II and older Nanopure systemTap or PretreatedOne 09-034 high capacity ion exchange, 09-034-104 Organic/colloid and two 09-034-3 Ultrapure™ ion exchange cartridgesD3804
Catalog number D3804
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
Nanopure II and older Nanopure system
Feedwater Source:
Tap or Pretreated
Includes:
One 09-034 high capacity ion exchange, 09-034-104 Organic/colloid and two 09-034-3 Ultrapure™ ion exchange cartridges
Model:
D3804
Choose from an array of Thermo Scientific™ Barnstead™ Cartridge Kits for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Specifications
Feedwater SourceTap or Pretreated
Flow Rate3.8
For Use With (Application)Water purification cartridge
For Use With (Equipment)Nanopure II and older Nanopure system
IncludesOne 09-034 high capacity ion exchange, 09-034-104 Organic/colloid and two 09-034-3 Ultrapure™ ion exchange cartridges
ModelD3804
Pressure30 psi
Shelf Life2 Years
TypeCartridge Kit
Length (English)17 in.
Length (Metric)4.18 cm
Diameter (English)3.38 in.
Diameter (Metric)8.5 cm
Unit SizeEA