E. HISTOLYTICA II™ Diluent
Product Image
E. HISTOLYTICA II™ Diluent
The E. HISTOLYTICA II™ test is an enzyme immunoassay for the rapid detection of the adhesin of E. histolytica in human fecal specimens. It is indicated for use with fecal specimens from patients with diarrhea or dysentery to determine the presence of E. histolytica gastrointestinal infection.
Have Questions?
Catalog NumberQuantity
D21 Ea.
Catalog number D2
Price (MYR)
8,406.00
EA
Add to cart
Quantity:
1 Ea.
Price (MYR)
8,406.00
EA
Add to cart
  • The test can be used for fecal specimens submitted for routine clinical testing from adults or children.
  • Conventional microscopy is not a prerequisite for use of the test.
Specifications
Product TypeDiluent
Quantity1 Ea.
CombinabilityNon-Combinable
Research AreaImmunology
Unit SizeEA
Frequently asked questions (FAQs)
Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.