Traceable™ Digital Thermometer
Actual product may vary
Traceable™ Digital Thermometer
The laboratory standard
Have Questions?
Catalog NumberTemperature Range (Metric)
CTL15-077-8-40°C to +150°C
Catalog number CTL15-077-8
Price (MYR)
3,540.00
Each
Temperature Range (Metric):
-40°C to +150°C
Price (MYR)
3,540.00
Each
Provides routine measurements with semisolid, liquid, or air/gas. Fisherbrand™ Traceable™ Digital Thermometers can switch between Celsius and Fahrenheit temperatures. Easy-to-read LCD screen includes Min/Max memory. Switch-selectable temperature ranges: -50° to +150°C and -58° to +302°F.
  • Universal, stainless-steel 316 probe is triple purpose for liquid, air/gas, and semisolids.
  • Ideal for foods, pharmaceuticals, petroleum products, gas systems, soil, ovens, and more
  • Unit reads in °C and °F and includes a HOLD key for freezing data on the display for future readings
  • Instant HISTORY key recalls minimum and maximum readings over the past 24 hours
  • Unit conforms to ITS-90 International Temperature Standard
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Depth (English)1.26 in.
Depth (Metric)32 mm
Display Dimensions0.5 in.H (13 mm)
For Use With (Application)Measuring temperatures in cuvettes, gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, petroleum products, foods and wastewater
Height (English)5.5 in.
Height (Metric)140 mm
IncludesStainless steel probe with USB connector; 9V Alkaline battery; High-impact, chemical-resistant ABS plastic case
Length (English) Cable59 in.
Length (English) Probe6 in.
Length (Metric)29 cm
Length (Metric) Cable150 cm
Length (Metric) Probe152.4 mm
MaterialABS Plastic
Probe TypeStainless Steel with USB Connector
Resolution0.0001°C
Weight (English)8 oz.
Weight (Metric)226.8 g
Width (English)3.5 in.
Product TypeDigital Thermometer
Length (English)9 in.
Max. Temperature (English)300°F
Max. Temperature (Metric)150°C
Min. Temperature (English)-40°F
Min. Temperature (Metric)-40°C
Temperature Range (English)-40°F to +300°F
Temperature Range (Metric)-40°C to +150°C
CalibratedYes
Accuracy±0.05°C
DisplayDigital LCD
Width (Metric)89 mm
Unit SizeEach