Traceable™ Nano™ Timer
Actual product may vary
Traceable™ Nano™ Timer
Offers ultra-small size with intuitive operation
Have Questions?
Catalog NumberTime Range
CTL14-649-8323 hr. 59 min. 59 sec.
Catalog number CTL14-649-83
Price (MYR)
131.00
Each
Time Range:
23 hr. 59 min. 59 sec.
Price (MYR)
131.00
Each
Offers ultra-small size with intuitive operation. Fisher Scientific™ Traceable™ Nano Timer displays countdown alarm time/stopwatch (countup) time, and time of day (AM/PM and 24-hour format).
  • Counts down or up to 23 hours, 59 minutes, 59 seconds on two timing channels
  • Accuracy: 0.01%
  • 0.375 in. high digits may be read from 10 ft. (3m)
  • For repetitive tests, two-channel memory returns display to original programmed time
Specifications
Product TypeHandheld/Benchtop Timer
Time Range23 hr. 59 min. 59 sec.
AlarmsHigh db alarm sounds for 1 min.
Accuracy0.0001
Resolution1 sec.
DisplayLCD
Dimensions (L x W x H)2.5 x 0.5 x 2 in. (6.4 x 1.3 x 5.08 cm)
Length (English)2.5 in.
Length (Metric)6.4 cm
Width (English)0.5 in.
Width (Metric)1.3 cm
Height (English)2 in.
Height (English) Display0.375 in.
Height (Metric)5.08 cm
Height (Metric) Display0.93 cm
Weight (English)2.5 oz.
Weight (Metric)71 g
IncludesTraceable certificate
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEach