Traceable™ Triple-Display Timer
Actual product may vary
Traceable™ Triple-Display Timer
Traceable™ Triple-Display Timer
Traceable™ Triple-Display Timer
Three-line display permits countdown of three tests simultaneously
Have Questions?
Catalog NumberTime Range
CTL06-662-319 hr. 59 min. 59 sec.
Catalog number CTL06-662-3
Price (MYR)
-
Time Range:
19 hr. 59 min. 59 sec.
Perfect timer for dedicating to a test requiring several different times
  • Get timing faster—recall all channels' settings instantly
  • Use as a countdown timer, count up stopwatch, or clock
  • Count up to 24 hours, count down to 20 hours
  • Displays time elapsed since alarm
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Product TypeHandheld/Benchtop Timer
Time Range19 hr. 59 min. 59 sec.
AlarmsAudible
Accuracy0.0001
Resolution1/100 sec. - Count-up mode; 1 sec. - Countdown Mode
DisplayDigital LCD
No. of Channels3
ColorBlack
Height (English) Display0.5 in.
Weight (English)3 oz.
Weight (Metric)0.09 kg
IncludesBattery, magnetic back, Traceable Certificate
Unit SizeEach