Traceable™ Timer
Traceable™ Timer
Actual product may vary
Traceable™ Timer
Traceable™ Timer
Traceable™ Timer
Four independent timing channels feature unique audible alarms for simultaneous timing
Have Questions?
Catalog NumberTime Range
CTL02-261-84099 hr. 59 min. 59 sec.
Catalog number CTL02-261-840
Price (MYR)
251.00
Each
Add to cart
Time Range:
99 hr. 59 min. 59 sec.
Price (MYR)
251.00
Each
Add to cart
Assorted bold colors for distinctive appearance. Fisher Scientific™ Traceable™ Timer is supplied with 3 unique colorful frames, permitting personalization.
  • Unique memory returns channel to previously programmed setting at touch of a key
  • Counts up during alarming indicating elapsed time since alarming, 12/24 hour clock
Count up or down from 100 hours for longer duration experiments. Memory function returns the display to the programmed time.
Traceable™ Products have an individually-numbered Traceable™ Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Specifications
Product TypeHandheld/Benchtop Timer
Time Range99 hr. 59 min. 59 sec.
AlarmsAudible
Accuracy0.0001
Resolution1 sec.
DisplayLCD
No. of Channels4
Battery TypeNo. 357 size silver oxide
ColorWhite
Weight (English)0.2 lb.
Weight (Metric)0.091 kg
IncludesBatteries, Easel Bench Stand, Magnetic Back
Certifications/ComplianceNIST
Unit SizeEach