15 mL Conical Centrifuge Tubes
Actual product may vary
15 mL Conical Centrifuge Tubes
Tested to withstand centrifugation of 12,000 RCF (polypropylene) or 1,800 RCF (polystyrene) in a fully supported rotor with room temperature water. Falcon™ 15mL Conical Centrifuge Tubes are ideal for cell centrifugation; pelleting; separation by density gradients.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackagingMaterialMax. RCF
CRG352097RackPolypropylene12,000 x g
CRG352096BulkPolypropylene12,000 x g
Catalog number CRG352097
Price (MYR)
664.00
Case
Packaging:
Rack
Material:
Polypropylene
Max. RCF:
12,000 x g
Price (MYR)
664.00
Case
  • Meet bioanalytical-grade requirements and provide unsurpassed performance in critical research applications
  • Unique double threaded cap design reducing cross threading and leakage
  • Easy-to-read graduations
  • Consistent biological and physical properties
  • Ideal for long-term cryogenic storage of specimens at -80°C (polypropylene tubes)
The most dependable tube in the lab
Falcon conical tubes are easy-to-use, high quality tools to protect your precious samples. State-of-the-art design and manufacturing create tubes engineered to provide high strength, wide temperature stability, and to perform in your critical applications. They will protect your valuable samples during centrifugation, vortexing, and long term storage in the freezer. To meet this intense challenge, Falcon tubes are designed for:
  • High strength: State-of-the-art mold design, coupled with advanced resin selection, create tube walls that are engineered to perform under high-stress situations
  • Nonpyrogenicity: Tested to less than 0.1 EU/mL
  • Nontoxicity: Resins are selected via an intense array of U.S. Pharmacopoeia (USP) toxicity tests
  • Low protein binding: Corning engineers and scientists are continually searching for materials and processes that minimize labware-induced interference, such as protein binding
  • Quality packaging: Falcon tubes, in addition to offering bioanalytical-grade performance, use medical-style packaging to better assure sterile presentation. The conical tubes are packaged in convenient reusable racks or compact environmentally-friendly bulk packs.
Specifications
Certifications/ComplianceISO 9001:2000, FDA, CFR
Closure MaterialPolyethylene
Closure ColorBlue
Diameter (English) Outside0.7 in.
ColorClear
Capacity (Metric)15 mL
MaterialPolypropylene
For Use WithCell centrifugation (pelleting bacterial cells, separation by density gradients); Molecular biology (purification and precipitation of nucleic acids), Sample storage (stable -80° to +121°C); Centrifugation of precipitates
No. per Case500
PackagingRack
SterilitySterile
ShapeConical
Length (Metric)120 mm
Capacity (English)0.507 oz.
Bottom ShapeConical
Closure TypeDome Seal Screw Cap
Writing AreaWhite Marking Spot
Diameter (Metric)17 mm
GraduatedYes
Graduations0.5 mL
Product TypeCentrifuge Tube
Max. RCF12,000 x g
Unit SizeCase