Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Actual product may vary
Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Arctic Express™ Dual Storage Systems
Store and ship samples with Arctic Express dual storage systems, which feature hard-shell shipping containers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
CK5092228 L
CK5092010 L
CK5092118.5 L
Catalog number CK50922
Price (MYR)
27,873.00
EA
Add to cart
Capacity (Metric):
28 L
Price (MYR)
27,873.00
EA
Add to cart
Protect nonhazardous samples during shipment and storage using Thermo Scientific™ Arctic Express™ Storage Systems. Featuring excellent flexibility and safety features, these systems can be used as dry shippers or laboratory cryo vessels.
Features:
  • When filled with liquid nitrogen and placed in the laboratory, unit can be used as a “can and cane” storage system indefinitely with static holding times of up to 125 days.
  • Aluminum vessels come with lockable lid and hold six numbered, color-coded, stainless-steel cans (included), which accommodate canes that hold ampules.
  • Warranty varies by country, please contact us for details
Specifications
TypeManual Fill LN2 Dewar
Capacity (Metric)28 L
No. of Doors1
ModelArctic Express Dual 28
Static Holding Time21 shipper/50 lab
Diameter (English)18.2 in.
Diameter (English) Inside Neck3.8 in.
Diameter (Metric)46.2 cm
Diameter (Metric) Inside Neck9.7 cm
Vial Capacity630 x 2mL
Unit SizeEA