Support Rod
Support Rod
Actual product may vary
Support Rod
For Use With: Isotemp hotplates, stirrers and stirring hotplates
Have Questions?
Catalog NumberType
CIC0001832Rod
Catalog number CIC0001832
Price (MYR)
146.00
EA
Add to cart
Type:
Rod
Price (MYR)
146.00
EA
Add to cart
Fisherbrand Hotplate/Stirrer Support Rod is to be used together with clamps and clamp holder to hold apparatus suitable for Isotemp hotplates, stirrers and stirring hotplates
  • Support rod
Specifications
For Use With (Equipment)Isotemp hotplates, stirrers and stirring hotplates
For Use With (Application)Use together with clamps and clamp holder to hold apparatus
TypeRod
Unit SizeEA