External Probe
External Probe
Actual product may vary
External Probe
For Use With: Isotemp (±1.8°C at 70°C) and Isotemp Advanced (±1.2°C at 70°C) Hotplates and Hotplate Stirrers
Have Questions?
Catalog NumberTemperature (Metric)
CIC00014650°C to 400°C
Catalog number CIC0001465
Price (MYR)
793.00
EA
Add to cart
Temperature (Metric):
0°C to 400°C
Price (MYR)
793.00
EA
Add to cart
  • Optimize performance and flexibility
Specifications
For Use With (Equipment)Isotemp (±1.8°C at 70°C) and Isotemp Advanced (±1.2°C at 70°C) Hotplates and Hotplate Stirrers
Temperature (Metric)0°C to 400°C
TypeProbe
Unit SizeEA