Choice™ Nylon Syringe Filters
Choice™ Nylon Syringe Filters
Actual product may vary
Choice™ Nylon Syringe Filters
Use Thermo Scientific™ Choice™ Nylon Syringe Filters as the economic option for most general laboratory filtration, and for filtration of most HPLC samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)
CH4525-NN0.45 μm25 mm
CH2225-NN0.2 μm25 mm
CH2213-NN0.2 μm13 mm
CH4513-NN0.45 μm13 mm
Catalog number CH4525-NN
Price (MYR)
-
Pore Size:
0.45 μm
Diameter (Metric):
25 mm
Thermo Scientific™ Choice™ Nylon Syringe Filters are a good option for most general laboratory filtration applications and filtration of most HPLC samples. These sryinge filters provide cleaner sample extracts by removing interfering materials and fine particles down to the rated pore size.
Adding a syringe filtration step prior to chromatographic injection not only helps to ensure more consistent and reliable results, it also helps protect delicate instruments and prolongs column life. Economic Choice Syringe Filters are available in a variety of sizes and membrane types to meet any application need.
Choice Nylon Syringe Filter Specifications
  • Membrane: HPLC certified nylon
  • Maximum operating temperature: 100 °C
  • Housing: Medical grade, virgin polypropylene
  • Autoclave: Sterilize by dry heat at 121 °C for 15 minutes
Choice Nylon Syringe Filter Applications
  • HPLC and organic-solvent sample preparation and clean-up
  • Dissolution sample analysis
  • General sample preparation
  • Mixed sample matrix of aqueous or organic dissolved analytes
Choice Nylon Syringe Filter Chemical Incompatibilities
  • Acids >1N
  • Halogenated solvents
  • Proteinaceous samples with high non-specific binding affinities
Specifications
DisposableSingle-use
Material (Membrane)Nylon
Pore Size0.45 μm
SterilityNon-sterile
For Use With (Application)HPLC, SPE
Housing MaterialSolvent Resistent, Low-Extractable Polypropylene
PrefilterNo
Pressure Rating90 psi
Product LineChoice
Quantity100/Pk.
TypeSyringe Filter
Diameter (Metric)25 mm
Unit SizePack