13mm Autosampler Vial Septa
13mm Autosampler Vial Septa
Actual product may vary
13mm Autosampler Vial Septa
13mm Septa for use with threaded caps Snap/Crimp Caps are available in PTFE/ Red Rubber, PTFE/Silicone or Red PTFE/White Silicone/Red PTFE.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorMaterialClosure SizeHardness
C4015-60Red, WhitePTFE, Silicone13-42550 °shore
C4015-10WhitePTFE13-42553 °shore
C4015-40
also known as TFS = C4015-40
Red, WhitePTFE, Silicone13-42555 °shore
C4015-45Red, WhitePTFE, Silicone13-42545 °shore
NSCC4015-60Red, WhitePTFE, Silicone13-42550 °shore
12-ST101BluePTFE, Silicone12 mm30 °shore
Catalog number C4015-60
Price (MYR)
-
Color:
Red, White
Material:
PTFE, Silicone
Closure Size:
13-425
Hardness:
50 °shore
For use with 13-425 threaded caps or 13mm Snap/Crimp Caps, Thermo Scientific™ 13mm Septa are available in PTFE/ Red Rubber, PTFE/Silicone or Red PTFE/White Silicone/Red PTFE to suit a variety of applications.
  • PTFE disk septa are chemically inert
  • PFTE/red rubber septa are economical for many GC applications
  • PTFE/silicone septa are most popular for HPLC and IC applications
  • Pre-slit septa are easier to pierce with thin gauge needles
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSeptum for 13-425 Screw Caps
ColorRed, White
MaterialPTFE, Silicone
Pre-slitYes
SeptumRed PTFE/White Silicone
Hardness50 °shore
Quantity100
Thickness (Metric)1.5 mm
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
TypeSepta
Closure Size13-425
For Use With (Equipment)13 to 425 Screw Cap
Unit SizePack