Screw Top Vial Racks
Screw Top Vial Racks
Actual product may vary
Screw Top Vial Racks
Efficiently hold and organize vials while using them or for storage. Save space by stacking the 50 or 100 plastic vial racks on top of each other.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsFor Use With (Equipment)Description
C4015-25Vials4 mL VialsVial Rack
FIS03-375-9Vials2 mL VialsVial Rack
Catalog number C4015-25
Price (MYR)
-
Holds:
Vials
For Use With (Equipment):
4 mL Vials
Description:
Vial Rack
Store vials efficiently with Thermo Scientific™ National 4mL Screw Top Vial Racks, stackable for even more space savings.
  • Racks feature alphanumeric indexing for easier vial identification
  • Polypropylene vial racks are resistant to most solvents
Ordering Information
50 and 100 vial capacity racks available.
Limited warranty period:
Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Product TypeVial Rack
DescriptionVial Rack
ColorClear
HoldsVials
MaterialPolypropylene
Positions50
ShapeSquare
StackableYes
For Use With (Equipment)4 mL Vials
Unit SizePack