11 mm Non-Assembled Clear Crimp Top & Snap-It Autosampler Vial Kits
11 mm Non-Assembled Clear Crimp Top & Snap-It Autosampler Vial Kits
Actual product may vary
11 mm Non-Assembled Clear Crimp Top & Snap-It Autosampler Vial Kits
Save time and reduce the risk of contamination using this convenience kit. Each kit includes 100 clear glass vials and 100 crimp top caps with pre-assembled septa.
Have Questions?
Catalog NumberMarking SpotClosure ColorClosure TypeSeptum
C4011-73NoClearSnap Cap, Open Top, 6 mm holePTFE/Silicone
Catalog number C4011-73
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Marking Spot:
No
Closure Color:
Clear
Closure Type:
Snap Cap, Open Top, 6 mm hole
Septum:
PTFE/Silicone
Save time during sample preparation and reduce the risk of contamination using Thermo Scientific™ 11 mm Crimp Top Clear Vial Convenience kits (Unassembled). Each kit includes 100 clear glass vials and 100 crimp top caps with pre-assembled septa. Reusable two-compartment trays protect vials and closures while keeping matching supplies together. Clear trays to make it easy to keep track of available supplies without opening containers. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Choose from a variety of autosampler vial kits.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorClear
DescriptionClear Glass; Septum: PTFE/Silicone
Diameter (Metric) Inner12 mm
ColorClear
MaterialGlass, PTFE, Silicone
Pre-slitNo
Volume (Metric)2 mL
SeptumPTFE/Silicone
IncludesVial: C4011-5, Closure: C4011-54
No. per Pack100 Pk.
Volume (Metric) Total2 mL
Closure MaterialPolyethylene
Closure TypeSnap Cap, Open Top, 6 mm hole
Height (Metric)32 mm
TypeUnassembled Snap Cap Vial Convenience Kit
Closure Size11 mm
Marking SpotNo
Septa Size (Metric)11 mm
Unit SizeBox