National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa
National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa
Actual product may vary
National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa
Thermo Scientific™ National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa include shrink-wrapped vials and separately packaged caps and septa.
Have Questions?
Catalog NumberVolume (Metric)Height (Metric)Diameter (Metric) Outer
B7990-3A8 mL60 mm17 mm
Catalog number B7990-3A
Price (MYR)
-
Volume (Metric):
8 mL
Height (Metric):
60 mm
Diameter (Metric) Outer:
17 mm
Thermo Scientific™ National Glass Screw-Thread Sample Vials with Open-Top Caps and Septa include shrink-wrapped vials and separately packaged caps and septa.
  • Unassembled convenience kits
  • 51A amber glass
  • Eliminate leaching of ions
  • Provide consistent pH for duration of sample storage life
  • With polypropylene open top screw caps and PTFE/silicone septa
  • Convenient packaging for storerooms or laboratory cabinets
Specifications
ColorAmber
Diameter (Metric) Outer17 mm
MaterialGlass
Volume (Metric)8 mL
SeptumPTFE/Silicone
Volume (English)2.125 dr.
Closure TypeScrew Cap (Open-top) with Septum
Height (Metric)60 mm
Closure Size15-425
Unit SizeBox